Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M6531

Sigma-Aldrich

15-Methylpalmitic acid

≥98% (capillary GC)

Synonym(s):

15-Methylhexadecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H34O2
CAS Number:
Molecular Weight:
270.45
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (capillary GC)

form

powder

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC(C)CCCCCCCCCCCCCC(O)=O

InChI

1S/C17H34O2/c1-16(2)14-12-10-8-6-4-3-5-7-9-11-13-15-17(18)19/h16H,3-15H2,1-2H3,(H,18,19)

InChI key

IIUXHTGBZYEGHI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyleann K Brooks et al.
PloS one, 4(10), e7545-e7545 (2009-10-22)
The nematode Caenorhabditis elegans has emerged as an important model for studies of the regulation of fat storage. C. elegans feed on bacteria, and various strains of E. coli are commonly used in research settings. However, it is not known
Atsuko Matsumoto et al.
International journal of systematic and evolutionary microbiology, 57(Pt 11), 2651-2654 (2007-11-06)
Two novel actinomycete strains, KV-744T and KV-856, were isolated by long-term cultivation. Aerial long-chain spores were produced directly from vegetative mycelia and possessed no motility. Vegetative mycelia developed very well and exhibited fragmentation. The cell-wall peptidoglycan contained meso-diaminopimelic acid, glycine
A A Aliero et al.
Journal of medicinal food, 10(3), 557-558 (2007-09-25)
The volatile components obtained by hydrodistillation of Solanum pseudocapsicum roots were analyzed by gas chromatography-mass spectrometry. A total of 41 compounds, representing 50% of the oil, were identified. The oil was found to contain fatty acids (26.8%), terpenoids (7.6%), and
Myung Soo Park et al.
International journal of systematic and evolutionary microbiology, 56(Pt 2), 433-438 (2006-02-02)
Two Gram-negative, yellow-pigmented bacteria designated PSD1-4T and PHA3-4T, isolated from two sand-dune plant species inhabiting coastal areas in Tae-an, Korea, were subjected to taxonomic investigation. 16S rRNA gene sequence analysis indicated that both isolates should be placed in the genus
Honoka Tanno et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1866(1), 158842-158842 (2020-10-19)
Although most mammalian fatty acids (FAs) are straight-chain, there also exist branched-chain FAs such as iso- and anteiso-FAs, especially in the meibomian glands. Meibum lipids, which are secreted from the meibomian glands and are important for dry eye prevention, contain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service