Skip to Content
MilliporeSigma
All Photos(1)

Documents

M6273

Sigma-Aldrich

2-O-Methylthymidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O5
CAS Number:
Molecular Weight:
256.26
UNSPSC Code:
12352204

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Singer et al.
Proceedings of the National Academy of Sciences of the United States of America, 80(16), 4884-4888 (1983-08-01)
O2-and O4-alkyldeoxythymidine are among the four O-alkyl base-modified derivatives produced by the reaction of N-nitroso alkylating agents with nucleic acids in vitro and in vivo. We find that both O2- and O4-methyl-dTTP can substitute for dTTP in alternating poly(dA-dT)-primed DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service