Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5698

Sigma-Aldrich

Anti-MIB2 (C-terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Mindbomb homolog 2, Anti-Novel zinc finger protein, Anti-Novelzin, Anti-Skeletrophin, Anti-Zinc finger ZZ type with ankyrin repeat domain protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~110 kDa

species reactivity

human, rat

technique(s)

western blot: 1:500-1:1,000 using lysates of rat skeletal muscle

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MIB2(142678)

General description

Mindbomb homolog 2 (MIB2) which is also known as skeletrophin, is an ubiquitin ligase. The gene encoding it is localized on human chromosome 1p36.32.
Mouse MIB1 and MIB2 share 52% similarity in their amino acid sequences and contain a similar domain organization.These include two homologous to the E6-AP carboxyl terminus (HECT) domains. It also comprises RCC1-like domain (RLD) domain containing E3 ubiquitin protein ligase 2 (herc2)/mib domains.Other domains associated with MIB2 are one ZZ zinc finger, one mib repeat, ankyrin repeats, and two interesting new gene (RING) domains. MIB2 is highly expressed in adult tissues.

Specificity

Anti-MIB2 (C-terminal) specifically recognizes human and rat MIB2.

Immunogen

synthetic peptide corresponding to amino acides 950-966 of human MIB2 conjugated to KLH. The corresponding sequence differs by one amino acid in mouse.

Application

Anti-MIB2 (C-terminal) antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Disruption of the MIB2 gene results in exencephaly and appears to be less essential in mouse development.
Mindbomb homolog 2 (MIB2) takes part in the ubiquitination of Notch ligands like Jagged-2 (JAG2). It binds to actin and CYLD (cylindromatosis (turban tumor syndrome)), which is an ubiquitin hydrolase. It has also been shown that MIB2 negatively regulates the transcription of Met oncogene.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiang I Wu et al.
Genesis (New York, N.Y. : 2000), 45(11), 722-727 (2007-11-08)
Mib1 and Mib2 ubiquitin ligases are very similar in their domain construction. They partake in the Notch signaling pathway by ubiquitinating the Notch receptors Delta and Jagged prior to endocytosis. We have created a targeted mutation of Mib2 and show
Katrin Domsch et al.
PloS one, 12(3), e0173733-e0173733 (2017-03-11)
The proper differentiation and maintenance of myofibers is fundamental to a functional musculature. Disruption of numerous mostly structural factors leads to perturbations of these processes. Among the limited number of known regulatory factors for these processes is Mind bomb2 (Mib2)
Bon-Kyoung Koo et al.
PloS one, 2(11), e1221-e1221 (2007-11-29)
The Notch signaling pathway is an evolutionarily conserved intercellular signaling module essential for cell fate specification that requires endocytosis of Notch ligands. Structurally distinct E3 ubiquitin ligases, Neuralized (Neur) and Mind bomb (Mib), cooperatively regulate the endocytosis of Notch ligands
Tamotsu Takeuchi et al.
DNA and cell biology, 24(5), 339-344 (2005-05-05)
Recent experiments have revealed that aberrant functionality of the chromatin remodeling complex is related to tumorigenicity in various malignant tumors. Skeletrophin is an actin-binding cytoskeleton-related molecule, which is induced by the overexpression of truncated human SWI1 (SMARCF1). Human SWI1 is
T Takeuchi et al.
Oncogene, 25(53), 7059-7069 (2006-05-23)
Skeletrophin (mindbomb homolog 2 (MIB2)) is a RING (Really Interesting New Gene) finger-dependent ubiquitin ligase, which targets the intracellular region of Notch ligands. A previous immunohistochemical study demonstrated that skeletrophin was downregulated in many melanomas. In the present study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service