Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5664

Sigma-Aldrich

4-Methylumbelliferyl-β-D-glucuronide hydrate

BioXtra

Synonym(s):

4-Methylumbelliferyl-β-D-glucopyranosiduronic acid, MUG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16O9 · xH2O
CAS Number:
Molecular Weight:
352.29 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

product line

BioXtra

Assay

≥98% (TLC)

form

powder

impurities

≤0.005% Phosphorus (P)

solubility

pyridine: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.05%

cation traces

Al: ≤0.001%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.001%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.001%

storage temp.

−20°C

SMILES string

O.CC1=CC(=O)Oc2cc(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(O)=O)ccc12

InChI

1S/C16H16O9.H2O/c1-6-4-10(17)24-9-5-7(2-3-8(6)9)23-16-13(20)11(18)12(19)14(25-16)15(21)22;/h2-5,11-14,16,18-20H,1H3,(H,21,22);1H2/t11-,12-,13+,14-,16+;/m0./s1

InChI key

URVSQZMOFUEQAW-YYHOVTOASA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service