Skip to Content
MilliporeSigma
All Photos(1)

Documents

M5567

Sigma-Aldrich

Anti-Munc-18 Interacting Protein 1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Mint1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~130 kDa

species reactivity

rat

technique(s)

western blot: 1 μg/mL using rat forebrain postnuclear supernatant

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Munc-18 interacting protein 1 (Mint1) is an adaptor protein primarily localized in the brain. It possesses phosphotyrosine-binding (PTB) domains and PDZ (PSD95, Dlg1 and zo-1) domains which are common to its family.

Specificity

Detects rat munc-18 interacting protein 1 (mint1) using rat brain extract and extract from HEK293 cells overexpressing the rat gene.

Immunogen

synthetic peptide corresponding to amino acid residues 1-17 from rat mint1.

Biochem/physiol Actions

Munc-18 interacting protein 1 (Mint1) binds to amyloid-β precursor protein (APP) and also associates with calcium/calmodulin-dependent serine protein kinase (CASK).

Physical form

Solution in phosphate buffered saline containing 1.0 mg/mL bovine serum albumin and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Biederer et al.
The Journal of biological chemistry, 275(51), 39803-39806 (2000-10-19)
Mint1 (X11/human Lin-10) and Mint2 are neuronal adaptor proteins that bind to Munc18-1 (n/rb-sec1), a protein essential for synaptic vesicle exocytosis. Mint1 has previously been characterized in a complex with CASK, another adaptor protein that in turn interacts with neurexins.
Wei Zhang et al.
Biochemical and biophysical research communications, 320(3), 717-721 (2004-07-09)
Munc-18-interacting (Mint) proteins are adaptors involved in regulation of synaptic vesicle exocytosis. We have investigated expression and cellular localization of Mint1 in pancreatic islets with special reference to insulin-secreting beta-cells. Western blotting showed that Mint1 was expressed in hamster (HIT-T15)
Thomas Biederer et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 22(17), 7340-7351 (2002-08-28)
Mints/X11s are neuron-specific (Mints 1 and 2) and ubiquitous (Mint 3) adaptor proteins composed of isoform-specific N-terminal sequences and common C-terminal phosphotyrosine-binding (PTB) and PDZ domains. We now show that all three Mints bind to the cytoplasmic tail of amyloid-beta

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service