Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5065

Sigma-Aldrich

MMP Control-5

buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

form

buffered aqueous solution

Quality Level

shipped in

dry ice

storage temp.

−20°C

General description

Concentrated, serum-free, cell culture media from cells containing Xenopus collagenase-4 (MMP-18).

Application

Used as a qualitative positive control in immunoblotting for MMP-18.

Physical form

Solution in 0.01 M phosphate buffered saline

Analysis Note

Recommended use for immunoblotting: mix 1:1 with 2X SDS-PAGE buffer and apply 20-40 μl per lane.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service