Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5064

Sigma-Aldrich

Rp-2′-O-Monobutyryladenosine 3′,5′-cyclic monophosphorothioate

≥97% (HPLC), solid

Synonym(s):

Rp-2′-O-MB-cAMPS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

≥97% (HPLC)

form

solid

mol wt

apparent mol wt 437.3 Da

packaging

vial of 5 μmol (approx. 2 mg)

storage temp.

−20°C

SMILES string

CCCC(=O)O[C@@H]1[C@@H]2OP(O)(=S)OC[C@H]2O[C@H]1n3cnc4c(N)ncnc34

InChI

1S/C14H18N5O6PS/c1-2-3-8(20)24-11-10-7(4-22-26(21,27)25-10)23-14(11)19-6-18-9-12(15)16-5-17-13(9)19/h5-7,10-11,14H,2-4H2,1H3,(H,21,27)(H2,15,16,17)/t7-,10-,11-,14-,26?/m1/s1

InChI key

XSAXBAAZBXKFFH-IDYJAONBSA-N

Application

Eliminates first messenger-stimulated phosphorylation by cyclic AMP-dependent protein kinase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U M Vischer et al.
Blood, 91(1), 118-127 (1998-02-07)
von Willebrand factor (vWF) is stored and released from endothelial secretory granules called Weibel-Palade (WP) bodies. Acute release can be induced by thrombin, histamine, and other mediators of thrombosis or inflammation. Their effect is thought to be mediated by an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service