Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5059

Sigma-Aldrich

Anti-Matrix Metalloproteinase-18 Hinge Region antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Xenopus collagenase, Anti-MMP-18, Anti-MMP-21-A, Anti-XMMP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen 53 kDa
antigen (pro-form) 51 kDa

species reactivity

Xenopus

concentration

~1 mg/mL

technique(s)

immunohistochemistry (frozen sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1:1,000

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Xenopus laevis ... mmp21-A(378572)

Specificity

Reacts with reduced and non-reduced MMP-18. Recognizes the pro-form as well as the smaller active forms of MMP-18.

Immunogen

synthetic peptide corresponding to the hinge region of Xenopus matrix metalloproteinase-18 (Xenopus collagenase, collagenase-4).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 50% glycerol and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service