Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5034

Sigma-Aldrich

Myristelaidic acid

~99% (capillary GC), liquid

Synonym(s):

trans-9-Tetradecenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26O2
CAS Number:
Molecular Weight:
226.36
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~99% (capillary GC)

form

liquid

storage temp.

−20°C

SMILES string

CCCC\C=C\CCCCCCCC(O)=O

InChI

1S/C14H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h5-6H,2-4,7-13H2,1H3,(H,15,16)/b6-5+

InChI key

YWWVWXASSLXJHU-AATRIKPKSA-N

Packaging

Sealed ampule.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service