Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5015

Sigma-Aldrich

Methyl Green

certified by the Biological Stain Commission

Synonym(s):

Ethyl Green

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C27H35BrClN3 · ZnCl2
CAS Number:
Molecular Weight:
653.24
Colour Index Number:
42590
Beilstein:
3914106
EC Number:
MDL number:
UNSPSC Code:
12171500

grade

certified by the Biological Stain Commission

composition

Dye content, ~90%

pH

0.2-1.8, yellow to blue

mp

>300 °C (lit.)

SMILES string

[Cl-].[Br-].Cl[Zn]Cl.CC[N+](C)(C)c1ccc(cc1)\C(c2ccc(cc2)N(C)C)=C3/C=C\C(C=C3)=[N+](\C)C

InChI

1S/C27H35N3.BrH.3ClH.Zn/c1-8-30(6,7)26-19-13-23(14-20-26)27(21-9-15-24(16-10-21)28(2)3)22-11-17-25(18-12-22)29(4)5;;;;;/h9-20H,8H2,1-7H3;4*1H;/q+2;;;;;+2/p-4

InChI key

RRKSQWOQZFIQKW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Quality

Material may be a mixture of monobromomonochloro and dichloro ZnCl2 salts. Halogen content listed on label.

Suitability

Certified for use as a counterstain in Altmann′s aniline acid fuchsin technique for mitochondria; for use in methyl green-pyronin methods for differential coloration of nucleic acids.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis and testing of biological stains--the Biological Stain Commission Procedures.
D P Penney et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 77(5-6), 237-275 (2003-02-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service