Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M4889

Sigma-Aldrich

9-Methyluric acid

powder

Synonym(s):

9-Methyl-2,6,8-trihydroxypurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O3
CAS Number:
Molecular Weight:
182.14
Beilstein:
18583
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

biological source

synthetic (organic)

form

powder

color

white to light yellow

SMILES string

CN1C(=O)NC2=C1NC(=O)NC2=O

InChI

1S/C6H6N4O3/c1-10-3-2(7-6(10)13)4(11)9-5(12)8-3/h1H3,(H,7,13)(H2,8,9,11,12)

InChI key

XJEJWDFDVPDMAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diane Yu et al.
RNA (New York, N.Y.), 26(8), 960-968 (2020-04-30)
Dozens of candidate orphan riboswitch classes have been discovered previously by using comparative sequence analysis algorithms to search bacterial genomic sequence databases. Each orphan is classified by the presence of distinct conserved nucleotide sequences and secondary structure features, and by
Gwen S Scott et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(25), 16303-16308 (2002-11-27)
Uric acid (UA) is a purine metabolite that selectively inhibits peroxynitrite-mediated reactions implicated in the pathogenesis of multiple sclerosis (MS) and other neurodegenerative diseases. Serum UA levels are inversely associated with the incidence of MS in humans because MS patients
Justyna Sekuła et al.
Analytica chimica acta, 875, 61-72 (2015-05-06)
Preparation is described of a durable surface of cationic gold nanoparticles (AuNPs), covering commercial and custom-made MALDI targets, along with characterization of the nanoparticle surface properties and examples of the use in MS analyses and MS imaging (IMS) of low

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service