Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M4750

Sigma-Aldrich

α-D(+)Mannose 1-phosphate bis(cyclohexylammonium) salt

Synonym(s):

α-D-Mannopyranosyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11O9P · 2C6H14N
CAS Number:
Molecular Weight:
458.48
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

>98% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

−20°C

SMILES string

NC1CCCCC1.NC2CCCCC2.OC[C@H]3O[C@H](OP(O)(O)=O)[C@@H](O)[C@@H](O)[C@@H]3O

InChI

1S/2C6H13N.C6H13O9P/c2*7-6-4-2-1-3-5-6;7-1-2-3(8)4(9)5(10)6(14-2)15-16(11,12)13/h2*6H,1-5,7H2;2-10H,1H2,(H2,11,12,13)/t;;2-,3-,4+,5+,6-/m..1/s1

InChI key

YBPDNWXTAIIEAM-SHPZNGBNSA-N

General description

Mannose is a monosaccharide.

Application

Mannose has been used in a study to assess the evolutionarily conserved glycan signal that degrades aberrant brassinosteroid receptors in Arabidopsis. It has also been used in a study to investigate preferences for uptake of carbohydrate-coated liposomes by C-type lectin receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi Hong et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(28), 11437-11442 (2012-06-27)
Asparagine-linked glycans (N-glycans) are crucial signals for protein folding, quality control, and endoplasmic reticulum (ER)-associated degradation (ERAD) in yeast and mammals. Although similar ERAD processes were reported in plants, little is known about their biochemical mechanisms, especially their relationships with
Yoko Kawauchi et al.
Glycoconjugate journal, 29(7), 481-490 (2012-06-27)
We evaluated the carbohydrate preferences of the C-type lectin receptors (CLRs) SIGNR1, SIGNR3, and Langerin as pathogen-uptake receptors based on uptake of liposomes consisting of cholesterol, DPPC, and various neoglycolipids at molar ratios of 10:10:1 and 10:7:4, respectively, using non-phagocytic
Silvia Guzman-Beltran et al.
Microbes and infection, 14(12), 1093-1101 (2012-07-10)
Sporothrix schenckii is a human pathogen that causes sporotrichosis, a cutaneous subacute or chronic mycosis. Little is known about the innate immune response and the receptors involved in host recognition and phagocytosis of S. schenckii. Here, we demonstrate that optimal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service