Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M4421

Sigma-Aldrich

5-(4-Methoxyphenyl)pyrrolo[2,1-d][1,5]benzothiazepin-4-ol N,N-dimethylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N2O3S
CAS Number:
Molecular Weight:
392.47
MDL number:
UNSPSC Code:
12352100

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C2=C(OC(=O)N(C)C)c3cccn3-c4ccccc4S2

Biochem/physiol Actions

Specific ligand for mitochondrial benzodiazepine receptors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Fiorini et al.
Journal of medicinal chemistry, 37(10), 1427-1438 (1994-05-13)
A novel class of ligands specific for MBR receptors has been identified: 6-arylpyrrolo[2,1-d][1,5]benzothiazepine derivatives. The majority of newly synthesized esters 37-64 as well as some intermediate ketones showed micro- or nanomolar affinity for [3H]PK 11195 binding inhibition. A SAR study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service