Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M3545

Sigma-Aldrich

Mastoparan from Polistes jadwagae

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C77H127N21O18
CAS Number:
Molecular Weight:
1634.96
UNSPSC Code:
41106300

Assay

≥97% (HPLC)

storage temp.

−20°C

Biochem/physiol Actions

Mast cell degranulating peptide. Activates phospholipase A2; inhibits calmodulin. Cell permeable; stimulates exocytosis and phosphoinositide breakdown.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Nakajima et al.
Peptides, 6 Suppl 3, 425-430 (1985-01-01)
In addition to wasp kinins, the wasp venom contains a series of hydrophobic peptides, mastoparans and chemotactic peptides as major peptidergic components. The first major component in the venom is mastoparam. The peptides in the mastoparan family are tetradecapeptide amides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service