Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M3316

Sigma-Aldrich

Myoseverin B

≥97% (HPLC)

Synonym(s):

2,6-Bis(4-methoxybenzylamino)-9-cyclohexylpurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H32N6O2
CAS Number:
Molecular Weight:
472.58
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

solubility

DMSO: soluble
H2O: insoluble

shipped in

dry ice

storage temp.

−20°C

SMILES string

COc1ccc(CNc2nc(NCc3ccc(OC)cc3)c4ncn(C5CCCCC5)c4n2)cc1

InChI

1S/C27H32N6O2/c1-34-22-12-8-19(9-13-22)16-28-25-24-26(33(18-30-24)21-6-4-3-5-7-21)32-27(31-25)29-17-20-10-14-23(35-2)15-11-20/h8-15,18,21H,3-7,16-17H2,1-2H3,(H2,28,29,31,32)

InChI key

ZUZXYJOSNSTJMU-UHFFFAOYSA-N

Biochem/physiol Actions

Myoseverin, a new microtubule-binding molecule, acts reversibly on myoblast proliferation without the cytotoxic effects displayed by nonpurine-based microtubule-disrupting molecules, like taxol, vinblastine, nocodazole. Myoseverin can be effectively used for the inhibition of new vessel growth by inhibiting endothelial cell function and differentiation of progenitor cells.
Tubulin polymerization inhibitor.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service