Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M3127

Sigma-Aldrich

S-Methylisothiourea hemisulfate salt

Synonym(s):

2-Methyl-2-thiopseudourea hemisulfate salt, S-Methyl-ITU, S-Methylthiuronium sulfate, SMT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN=C(NH2)SCH3 · 1/2H2SO4
CAS Number:
Molecular Weight:
139.19
Beilstein:
3717365
MDL number:
UNSPSC Code:
12352204

mp

240-241 °C (dec.) (lit.)

SMILES string

CSC(N)=N.CSC(N)=N.OS(O)(=O)=O

InChI

1S/2C2H6N2S.H2O4S/c2*1-5-2(3)4;1-5(2,3)4/h2*1H3,(H3,3,4);(H2,1,2,3,4)

InChI key

BZZXQZOBAUXLHZ-UHFFFAOYSA-N

Gene Information

human ... NOS2(4843)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

More potent than NMMA as an inhibitor of inducible nitric oxide synthetase (iNOS).

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - Solvent

Flash Point(C)

87 °C - Solvent


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service