Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

M2918

Sigma-Aldrich

2-Methoxy-4-(2-nitrovinyl)phenyl β-D-glucopyranoside

≥95%

Synonym(s):

MNP-Glc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19NO9
CAS Number:
Molecular Weight:
357.31
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95%

storage temp.

2-8°C

SMILES string

COc1cc(ccc1OC2OC(CO)C(O)C(O)C2O)\C=C/N(=O)=O

InChI

1S/C15H19NO9/c1-23-10-6-8(4-5-16(21)22)2-3-9(10)24-15-14(20)13(19)12(18)11(7-17)25-15/h2-6,11-15,17-20H,7H2,1H3/b5-4-

InChI key

VLHYYNTWHBRKNT-PLNGDYQASA-N

Substrates

Chromogenic substrate (505 nm) for β-glucosidase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service