Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M2769

Sigma-Aldrich

N-Methylformamide

Synonym(s):

N-Formylmethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONHCH3
CAS Number:
Molecular Weight:
59.07
Beilstein:
1098352
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

refractive index

n20/D 1.432 (lit.)

bp

198-199 °C (lit.)

mp

−4 °C (lit.)

density

1.011 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)NC

InChI

1S/C2H5NO/c1-3-2-4/h2H,1H3,(H,3,4)

InChI key

ATHHXGZTWNVVOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

231.8 °F - closed cup

Flash Point(C)

111 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service