Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M221

Sigma-Aldrich

Monoclonal Anti-Nicotinic Acetylcholine Receptor (α3 Subunit) antibody produced in rat

~5 mg/mL, clone mAb 313, affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

mAb 313, monoclonal

form

buffered aqueous solution

species reactivity

chicken

concentration

~5 mg/mL

technique(s)

immunohistochemistry (frozen sections): 1:4,000

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

chicken ... CHRNA3(386578)

Specificity

Binds to native and denatured detergent-solubilized α3 subunit of AchR. Does not bind α1, α4, α5, α7, α8 and β2.

Immunogen

recombinant large cytoplasmic domain of the α3 subunit (amino acids 315-441) of the nicotinic acetylcholine receptor from chick.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Solution in 20 mM Na3PO4, pH 7.2, containing 150 mM NaCl and 0.05% sodium azide.

Legal Information

Sold with permission of the Salk Institute

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W G Conroy et al.
Neuron, 9(4), 679-691 (1992-10-01)
The acetylcholine receptor (AChR) alpha 5 gene has been classified as a member of the AChR gene family based on sequence homology. Expression studies, however, have yet to identify a function for the alpha 5 gene product or even to
P J Whiting et al.
Brain research. Molecular brain research, 10(1), 61-70 (1991-04-01)
Neuronal nicotinic acetylcholine receptors (AChRs) are composed of two types of subunits: ACh-binding (termed alpha 2, alpha 3, alpha 4 ...) and structural (termed beta 2, beta 3, beta 4 ...). AChR subtypes composed of combinations of subunits of these
Lindstrom
Neurotransmissions, 12, 2-2 (1996)
Margarita Villar et al.
Biomolecules, 10(4) (2020-04-09)
Ticks are obligate hematophagous arthropods and vectors of pathogens affecting human and animal health worldwide. Cement is a complex protein polymerization substance secreted by ticks with antimicrobial properties and a possible role in host attachment, sealing the feeding lesion, facilitating
Benjaporn Buranrat et al.
Oncology reports, 34(5), 2790-2796 (2015-09-10)
Ferritin is a major iron storage protein and essential for iron homeostasis. It has a wide range of functions in the body including iron delivery, immunosuppression, angiogenesis, and cell proliferation. Ferritin is overexpressed in many cancer cells, but its precise

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service