Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M216

Sigma-Aldrich

MDL 105,519

≥98% (HPLC), solid

Synonym(s):

(E)-4,6-Dichloro-3-(2-phenyl-2-carboxyethenyl)indole-2-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$878.00
25 MG
$3,530.00

$878.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$878.00
25 MG
$3,530.00

About This Item

Empirical Formula (Hill Notation):
C18H11Cl2NO4
CAS Number:
Molecular Weight:
376.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

$878.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: >20 mg/mL

SMILES string

OC(=O)C(=C/c1c([nH]c2cc(Cl)cc(Cl)c12)C(O)=O)\c3ccccc3

InChI

1S/C18H11Cl2NO4/c19-10-6-13(20)15-12(16(18(24)25)21-14(15)7-10)8-11(17(22)23)9-4-2-1-3-5-9/h1-8,21H,(H,22,23)(H,24,25)/b11-8-

InChI key

LPWVUDLZUVBQGP-FLIBITNWSA-N

Gene Information

human ... GRIN1(2902)
rat ... Grin1(24408)

Biochem/physiol Actions

MDL 105,519 is a high affinity N-methyl-D-aspartate (NMDA) glutamate receptor antagonist at the glycine site. It inhibits NMDA-dependent responses.[1]

Legal Information

Sold under exclusive license from Hoechst Marion Roussel, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B M Baron et al.
European journal of pharmacology, 323(2-3), 181-192 (1997-04-04)
MDL 105,519, (E)-3-(2-phenyl-2-carboxyethenyl)-4,6-dichloro-1 H-indole-2-carboxylic acid, is a potent and selective inhibitor of [3H]glycine binding to the NMDA receptor. MDL 105,519 inhibits NMDA (N-methyl-D-aspartate)-dependent responses including elevations of [3H]N-[1,(2-thienyl)cyclohexyl]-piperidine ([3H]TCP) binding in brain membranes, cyclic GMP accumulation in brain slices, and
Pharmacological characterization of MDL 105,519, an NMDA receptor glycine site antagonist.
Baron BM
European Journal of Pharmacology, 323(2-3), 181-192 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service