Skip to Content
MilliporeSigma
All Photos(1)

Documents

M1816

Sigma-Aldrich

Myelin Basic Protein Kinase 1 from bovine kidney

>90% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
eCl@ss:
32160410
NACRES:
NA.32

Quality Level

Assay

>90% (SDS-PAGE)

form

solution

specific activity

≥300 units/mg protein

mol wt

40 kDa

shipped in

dry ice

storage temp.

−70°C

Unit Definition

One unit will incorporate one nanomole of phosphate into myelin basic protein at pH 7.0 at 30 °C

Physical form

Solution at 1 μg/mL in 50 mM Tris-HCl, pH 7.0, 1 mM benzamidine, 1 mM EDTA, 0.1 mM PMSF, 14 mM β-mercaptoethanol, and 50% glycerol

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service