Skip to Content
MilliporeSigma
All Photos(1)

Documents

M1257

Sigma-Aldrich

2-Mercaptopyridine

Synonym(s):

2-Pyridinethiol, 2-Pyridyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NS
CAS Number:
Molecular Weight:
111.16
Beilstein:
105787
EC Number:
MDL number:
UNSPSC Code:
12352204

mp

127-130 °C (lit.)

storage temp.

2-8°C

SMILES string

Sc1ccccn1

InChI

1S/C5H5NS/c7-5-3-1-2-4-6-5/h1-4H,(H,6,7)

InChI key

WHMDPDGBKYUEMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service