Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M1198

Sigma-Aldrich

MEK2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MAP2K2, MAPKK2, MKK2, PRKMK2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$593.00

$593.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$593.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$593.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

190-258 nmol/min·mg

mol wt

~71 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAP2K2(5605)

Biochem/physiol Actions

MEK2 is the member of MAPK kinase (MAPKK) family of signaling protein kinases. MEK2 is a dual-specificity kinase that activates the extracellular signal-regulated kinase (ERK) and mitogen-activated protein (MAP) kinase upon agonist binding to receptors. MEK2 plays a key role in the Ras/Raf/MEK/ERK mitogen-activated protein kinase (MAPK) signaling pathways. Approximately 30% of all human cancers have a constitutively activated MAPK pathway, and constitutive activation of MEK2 results in cellular transformation. The ERK/MAP kinase cascade regulates cell growth and differentiation.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Xu et al.
Molecular endocrinology (Baltimore, Md.), 11(11), 1618-1625 (1997-11-05)
Mitogen-activated protein (MAP)/ERK kinase (MEK)1 and MEK2 are the upstream activators of the MAP kinases, ERK1 and ERK2. MEK1 and MEK2 are approximately 85% identical in sequence but have unique inserts in their C-terminal domains. MEK isoform-specific antibodies were used
Louis-François Bélanger et al.
Molecular and cellular biology, 23(14), 4778-4787 (2003-07-02)
MEK is a dual-specificity kinase that activates the extracellular signal-regulated kinase (ERK) mitogen-activated protein (MAP) kinase upon agonist binding to receptors. The ERK/MAP kinase cascade is involved in cell fate determination in many organisms. In mammals, this pathway is proposed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service