Skip to Content
MilliporeSigma
All Photos(1)

Documents

M1032

Sigma-Aldrich

Cyclohexyl methacrylate

>97% (GC)

Synonym(s):

Methacrylic acid cyclohexyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COOC6H11
CAS Number:
Molecular Weight:
168.23
Beilstein:
2045235
EC Number:
MDL number:
UNSPSC Code:
12162002

Assay

>97% (GC)

contains

0.006% hydroquinone monomethyl ether as inhibitor

refractive index

n20/D 1.458 (lit.)

bp

68-70 °C/4 mmHg (lit.)

density

0.964 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OC1CCCCC1

InChI

1S/C10H16O2/c1-8(2)10(11)12-9-6-4-3-5-7-9/h9H,1,3-7H2,2H3

InChI key

OIWOHHBRDFKZNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

181.4 °F

Flash Point(C)

83 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service