Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

M0895

Sigma-Aldrich

MES potassium salt

≥99% (titration)

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid potassium salt, 4-Morpholineethanesulfonic acid potassium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$223.00

$223.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$223.00

About This Item

Empirical Formula (Hill Notation):
C6H12KNO4S
CAS Number:
Molecular Weight:
233.33
MDL number:
UNSPSC Code:
12161700
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
NA.25

$223.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥99% (titration)

form

crystalline powder

useful pH range

5.5-6.7

pKa 

6.1

solubility

water: 0.33 g/mL, clear, colorless

SMILES string

[K+].[O-]S(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S.K/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);/q;+1/p-1

InChI key

IMFIKFZWLAWMQE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

MES potassium salt has been used as a component of buffers for the preparation of permeabilized fiber bundles.[1][2]

Biochem/physiol Actions

MES is applicable as a Good′s buffer and is widely used in regulating the pH of reagent solutions, physiological experiments and plant culture medium.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoko Kagenishi et al.
Frontiers in plant science, 7, 79-79 (2016-03-01)
In plants, growth of roots and root hairs is regulated by the fine cellular control of pH and reactive oxygen species (ROS). MES, 2-(N-morpholino)ethanesulfonic acid as one of the Good's buffers has broadly been used for buffering medium, and it
Cody D Smith et al.
The Journal of biological chemistry, 295(48), 16207-16216 (2020-08-05)
Compensatory changes in energy expenditure occur in response to positive and negative energy balance, but the underlying mechanism remains unclear. Under low energy demand, the mitochondrial electron transport system is particularly sensitive to added energy supply (i.e. reductive stress), which
Ashley S Williams et al.
Cell metabolism, 31(1), 131-147 (2019-12-10)
This study sought to examine the functional significance of mitochondrial protein acetylation using a double knockout (DKO) mouse model harboring muscle-specific deficits in acetyl-CoA buffering and lysine deacetylation, due to genetic ablation of carnitine acetyltransferase and Sirtuin 3, respectively. DKO
Trace Thome et al.
American journal of physiology. Cell physiology, 317(4), C701-C713 (2019-07-11)
Chronic kidney disease (CKD) leads to increased skeletal muscle fatigue, weakness, and atrophy. Previous work has implicated mitochondria within the skeletal muscle as a mediator of muscle dysfunction in CKD; however, the mechanisms underlying mitochondrial dysfunction in CKD are not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service