Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M0640

Sigma-Aldrich

6-Methoxy-2-benzoxazolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NO3
CAS Number:
Molecular Weight:
165.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

color

light tan

mp

151-156 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccc2NC(=O)Oc2c1

InChI

1S/C8H7NO3/c1-11-5-2-3-6-7(4-5)12-8(10)9-6/h2-4H,1H3,(H,9,10)

InChI key

MKMCJLMBVKHUMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvejg K Mathiassen et al.
Journal of agricultural and food chemistry, 54(4), 1058-1063 (2006-02-16)
The hydroxamic acid 2,4-dihydroxy-7-methoxy-1,4-benzoxazin-3-one (DIMBOA) and the benzoxazolinones benzoxazolin-2-one (BOA) and 6-methoxybenzoxazolin-2-one (MBOA) have been identified as important allelochemicals in wheat. This study examines the possibility of exploiting the allelopathic properties of wheat as a weed control strategy by cultivating
Hisashi Kato-Noguchi et al.
Journal of chemical ecology, 32(5), 1101-1109 (2006-06-02)
6-Methoxy-benzoxazolin-2(3H)-one (MBOA) inhibited the germination of cress (Lepidium sativum L.) seeds at concentrations greater than 0.03 mM. Inhibition was overcome by sucrose, suggesting that MBOA may inhibit sugar metabolism in cress seeds. Induction of alpha-amylase activity in seeds was also
Tamara Coja et al.
Journal of agricultural and food chemistry, 54(4), 1093-1098 (2006-02-16)
Seven selected benzoxazinoid allelochemicals and synthetic reference compounds were tested for their lethal and sublethal effects in different field soils and standard soil on Folsomia candida and Poecilus cupreus by applying standard laboratory test procedures. The higher microbial activity in
S T Willard et al.
Animal reproduction science, 93(3-4), 280-291 (2005-09-13)
The objective of this investigation was to examine the effects of 6-methoxy-benzoxazolinone (MBOA), a plant compound that resembles melatonin and alters ovarian function in rodents, in combination with PMSG on superovulatory responses in the cycling ewe. In Experiment I, St.
M Fallas-López et al.
Animal reproduction science, 129(3-4), 221-228 (2012-01-25)
The sprouted wheat (SW) contains the 6-methoxy-2-benzoxazolinone (6-MBOA), a phenol compound that stimulates reproduction in certain small wild herbivorous mammals. The objective of the present study was to evaluate the effect of short-term supplemental dietary SW on libido, semen and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service