Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M0625

Sigma-Aldrich

Maleic anhydride

95% (may contain up to 5% maleic acid)

Synonym(s):

2,5-Furandione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2O3
CAS Number:
Molecular Weight:
98.06
Beilstein:
106909
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor density

3.4 (vs air)

vapor pressure

0.16 mmHg ( 20 °C)

Assay

95% (may contain up to 5% maleic acid)
99.02%

form

solid

autoignition temp.

870 °F

expl. lim.

7.1 %

bp

200 °C (lit.)

mp

51-56 °C (lit.)

SMILES string

O=C1OC(=O)C=C1

InChI

1S/C4H2O3/c5-3-1-2-4(6)7-3/h1-2H

InChI key

FPYJFEHAWHCUMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1A - STOT RE 1 Inhalation

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emmanuel J Favaloro et al.
Thrombosis research, 134(6), 1292-1300 (2014-10-11)
von Willebrand disease (VWD) is reportedly the most common bleeding disorder and arises from deficiency and/or defects of von Willebrand factor (VWF). Laboratory diagnosis and typing has important management implications and requires a wide range of tests, including VWF activity
Veronica H Flood et al.
Blood, 125(14), 2297-2304 (2015-02-11)
Von Willebrand factor (VWF) contains binding sites for platelets and for vascular collagens to facilitate clot formation at sites of injury. Although previous work has shown that VWF can bind type IV collagen (collagen 4), little characterization of this interaction
Ana Martinez et al.
Journal of medicinal chemistry, 48(23), 7103-7112 (2005-11-11)
The 2,4-disubstituted thiadiazolidinones (TDZD) are described as the first ATP-noncompetitive GSK-3 inhibitors. Following an SAR study about TDZD, different structural modifications in the heterocyclic ring aimed to test the influence of each heteroatom on the biological study are here reported
Glenn B S Miller et al.
European journal of mass spectrometry (Chichester, England), 21(3), 545-556 (2015-08-27)
We have conducted collision induced dissociation experiments on the hydrogen maleate anion (MaH(-), m/z = 115) and the anionic maleate MgCl complex (MaMgC(-), m/z = 173). In addition, we have computationally investigated the observed fragmentation reactions. We find that both
Neha N Parayath et al.
International journal of nanomedicine, 10, 4653-4667 (2015-08-01)
Drug delivery systems could potentially overcome low bioavailability and gastrointestinal toxicity, which are the major challenges for the development of oral anticancer drugs. Herein, we demonstrate the ability of styrene maleic acid (SMA) nanomicelles encapsulating epirubicin to traverse in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service