Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M0571

Sigma-Aldrich

MBP (85-99) peptide antagonist trifluoroacetate salt

95% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H114N18O21 · xC2HF3O2
CAS Number:
Molecular Weight:
1543.76 (free base basis)
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

95% (HPLC)

form

lyophilized powder

composition

Peptide content, 63% HPLC

solubility

H2O: >1 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(C)[C@H](NC(=O)[C@H](CCCCN)NC(=O)[C@@H]1CCCN1C(=O)[C@H](CCCCN)NC(=O)[C@@H](N)CCC(O)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc2ccc(O)cc2)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N3CCC[C@H]3C(=O)N[C@@H](C)C(O)=O

InChI

1S/C70H114N18O21.C2HF3O2/c1-36(2)55(86-63(101)47(18-10-13-31-72)82-66(104)52-21-16-34-88(52)69(107)49(19-11-14-32-73)84-60(98)45(74)26-28-53(90)91)67(105)83-48(27-29-54(92)93)62(100)78-40(6)59(97)85-50(35-43-22-24-44(89)25-23-43)64(102)81-46(17-9-12-30-71)61(99)77-39(5)57(95)75-37(3)56(94)76-38(4)58(96)79-41(7)68(106)87-33-15-20-51(87)65(103)80-42(8)70(108)109;3-2(4,5)1(6)7/h22-25,36-42,45-52,55,89H,9-21,26-35,71-74H2,1-8H3,(H,75,95)(H,76,94)(H,77,99)(H,78,100)(H,79,96)(H,80,103)(H,81,102)(H,82,104)(H,83,105)(H,84,98)(H,85,97)(H,86,101)(H,90,91)(H,92,93)(H,108,109);(H,6,7)/t37-,38-,39-,40-,41-,42-,45-,46-,47-,48-,49-,50-,51-,52-,55-;/m0./s1

InChI key

JHPGEXWNFDTTLX-RMOYKTNBSA-N

Amino Acid Sequence

Glu-Lys-Pro-Lys-Val-Glu-Ala-Tyr-Lys-Ala- Ala- Ala- Ala-Pro-Ala

Biochem/physiol Actions

MBP (85-99) peptide antagonist inhibits the binding of MBP 85-99 to HLA-DR2 and IL-2 and is a tool for MS research. The peptide antagonist is a 15-mer peptide that competes with Myelin Basic Protein MBP (85-99), the immunodominant epitope of MBP and major candidate autoantigen in multiple sclerosis (MS).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service