Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M0563

Sigma-Aldrich

[Met210]-Melanocyte Protein PMEL 17 Fragment 209-217 human, mouse

>98% (HPLC), solid

Synonym(s):

[Met53]-Melanocyte Protein PMEL 17 fragment 52-60 bovine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H74N10O14S
Molecular Weight:
1035.21
MDL number:
UNSPSC Code:
51111800

Assay

>98% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](N)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](C(C)C)C(=O)N1CCC[C@H]1C(=O)N[C@@H](Cc2ccccc2)C(=O)N[C@@H](CO)C(=O)N[C@@H](C(C)C)C(O)=O

Gene Information

human ... SILV(6490)

Amino Acid Sequence

Ile-Met-Asp-Gln-Val-Pro-Phe-Ser-Val

Biochem/physiol Actions

Modification of the melanoma differentiation antigen gp100/PMEL 17 fragment 209-217 sequence with greater affinity for the HLA-A2 molecule than the unmodified peptide. Has an increased ability to generate melanoma-reactive cytotoxic T lymphocytes in vitro.
Sequence with greater affinity for the HLA-A2 molecule than the unmodified peptide; has an increased ability to generate melanoma-reactive cytotoxic T lymphocytes in vitro.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service