Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M0546

Sigma-Aldrich

7-Methoxycoumarin-4-acetyl-Pro-Leu

≥97% (TLC)

Synonym(s):

MOCAc-Pro-Leu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28N2O7
CAS Number:
Molecular Weight:
444.48
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (TLC)

storage temp.

−20°C

SMILES string

COc1ccc2C(CC(=O)N3CCC[C@H]3C(=O)N[C@@H](CC(C)C)C(O)=O)=CC(=O)Oc2c1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C G Knight et al.
FEBS letters, 296(3), 263-266 (1992-01-27)
(7-methoxycoumarin-4-yl)Acetyl-Pro-Leu-Gly-Leu-(3-[2,4-dinitrophenyl]-L- 2,3-diaminopropionyl)-Ala-Arg-NH2 (Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH2) has been synthesised as a fluorogenic substrate for the matrix metalloproteinases. The highly fluorescent 7-methoxycoumarin group is efficiently quenched by energy transfer to the 2,4-dinitrophenyl group. The punctuated metalloproteinase (PUMP, EC 3.4.24.23) cleaves the substrate at the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service