Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M0449

Sigma-Aldrich

MEKK3, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MAP3K3, MAPKKK3

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$493.00

$493.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$493.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

$493.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

17-23 nmol/min·mg

mol wt

~105 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAP3K3(4215)

Biochem/physiol Actions

MEKK3 is a member of the MEKK family of protein kinases that directly regulate SAPK and ERK pathway but not the p38 pathway. MEKK3 modulates SAPK and ERK activity by activating the upstream targets SEK and MEK1/2, respectively. KSR2 can regulate the activity of MEKK3 and can reduce MEKK3-mediated ERK activation. Knockout of the MEKK3 gene in mice leads to disruption of blood vessel development and the structural integrity of the yolk sac. The blockage of angiogenesis in these mice is not a result of decreased VEGF1 expression but rather due to an intrinsic defect in Mekk3 -/- endothelial cells. Thus, MEKK3 is necessary for blood vessel development and may be a possible target for drugs that control angiogenesis.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Ellinger-Ziegelbauer et al.
The Journal of biological chemistry, 272(5), 2668-2674 (1997-01-31)
The extracellular signal-regulated kinase (ERK) pathway, the stress-activated protein kinase (SAPK) pathway, and the p38 pathway are three major mitogen-activated protein kinase (MAPK) cascades known to participate in the regulation of cellular responses to a variety of extracellular signals. Upstream

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service