Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L9905

Sigma-Aldrich

Lys-Arg-Thr-Leu-Arg-Arg trifluoroacetate salt

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H68N16O8
CAS Number:
Molecular Weight:
829.01
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

form

solid

composition

Peptide content, ≥60%

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(NC(=O)C(CCCNC(N)=N)NC(=O)C(N)CCCCN)C(C)O)C(=O)NC(CCCNC(N)=N)C(=O)NC(CCCNC(N)=N)C(O)=O

InChI

1S/C34H68N16O8/c1-18(2)17-24(29(55)47-21(10-6-14-43-32(37)38)27(53)48-23(31(57)58)12-8-16-45-34(41)42)49-30(56)25(19(3)51)50-28(54)22(11-7-15-44-33(39)40)46-26(52)20(36)9-4-5-13-35/h18-25,51H,4-17,35-36H2,1-3H3,(H,46,52)(H,47,55)(H,48,53)(H,49,56)(H,50,54)(H,57,58)(H4,37,38,43)(H4,39,40,44)(H4,41,42,45)

InChI key

QUJRRXNRGVULBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Protein kinase C substrate derived from EGF receptor

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service