Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L9628

Sigma-Aldrich

α-Lactose 1-phosphate barium salt

powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H21O14PBa
CAS Number:
Molecular Weight:
557.59
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

form

powder

color

white

storage temp.

−20°C

SMILES string

[Ba].OCC1OC(OC2C(O)C(O)C(OC2CO)OP(O)(O)=O)C(O)C(O)C1O

InChI

1S/C12H23O14P.Ba.2H/c13-1-3-5(15)6(16)8(18)11(23-3)25-10-4(2-14)24-12(9(19)7(10)17)26-27(20,21)22;;;/h3-19H,1-2H2,(H2,20,21,22);;;

InChI key

TZQWTVMDQLZZNA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service