All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H21N3O3
CAS Number:
Molecular Weight:
243.30
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.
storage temp.
−20°C
SMILES string
NCCCCC(N)C(=O)N1CCCC1C(O)=O
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service