Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L8886

Sigma-Aldrich

[des-Gly10, D-Phe6]-LH-RH ethylamide

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H82N16O12
CAS Number:
Molecular Weight:
1243.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CCNC(=O)C1CCCN1C(=O)C(CCCNC(N)=N)NC(=O)C(CC(C)C)NC(=O)C(Cc2ccccc2)NC(=O)C(Cc3ccc(O)cc3)NC(=O)C(CO)NC(=O)C(Cc4c[nH]c5ccccc45)NC(=O)C(Cc6cnc[nH]6)NC(=O)C7CCC(=O)N7

InChI

1S/C62H82N16O12/c1-4-66-60(89)51-17-11-25-78(51)61(90)44(16-10-24-67-62(63)64)71-54(83)45(26-35(2)3)72-55(84)46(27-36-12-6-5-7-13-36)73-56(85)47(28-37-18-20-40(80)21-19-37)74-59(88)50(33-79)77-57(86)48(29-38-31-68-42-15-9-8-14-41(38)42)75-58(87)49(30-39-32-65-34-69-39)76-53(82)43-22-23-52(81)70-43/h5-9,12-15,18-21,31-32,34-35,43-51,68,79-80H,4,10-11,16-17,22-30,33H2,1-3H3,(H,65,69)(H,66,89)(H,70,81)(H,71,83)(H,72,84)(H,73,85)(H,74,88)(H,75,87)(H,76,82)(H,77,86)(H4,63,64,67)

InChI key

DCHIRKIHIUVOFN-UHFFFAOYSA-N

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service