Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L8510

Sigma-Aldrich

(+)-Longifolene

Synonym(s):

(1R,2S,7S,9S)-3,3,7-Trimethyl-8-methylenetricyclo[5.4.0.02.9]undecane, Junipene, Kuromatsuene, [1S-(1α,3aβ,4α,8aβ)]-Decahydro-4,8,8-trimethyl-9-methylene-1,4-methanoazulene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
2044263
EC Number:
MDL number:
UNSPSC Code:
12352200

refractive index

n20/D 1.504 (lit.)

bp

254 °C/706 mmHg (lit.)

density

0.928 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)CCC[C@]2(C)C3CCC(C13)C2=C

InChI

1S/C15H24/c1-10-11-6-7-12-13(11)14(2,3)8-5-9-15(10,12)4/h11-13H,1,5-9H2,2-4H3/t11?,12?,13?,15-/m0/s1

InChI key

PDSNLYSELAIEBU-WOFVOEOOSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service