Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L7886

Sigma-Aldrich

Lectin from Ricinus communis (castor bean) Agglutinin RCA120

buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

form

buffered aqueous solution

potency

<4 μg/mL agglutination activity

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Lectins are carbohydrate binding proteins.[1] Lectin from Ricinus communis (castor bean) Agglutinin RCA120 is purified from the seeds of the castor plant. It is a tetrameric hemagglutinin with two α and two β subunits.[2]

Application

Lectin from Ricinus communis (castor bean) Agglutinin RCA120 has been used for the digestion, enrichment and deglycosylation of chicken egg white (CEW) glycopeptides.[3] It has also been used as a model carbohydrate-binding protein.[4]

Biochem/physiol Actions

Ricinus communis agglutinin (RCA) occurs in two forms designated RCA60 and RCA120 according to their mol. wt. of approx. 60,000 and 120,000 respectively. Neither RCA60 nor RCA120 is blood group specific. RCA60, also referred to as RCAII, Ricin D or RCL III is extremely toxic, inhibits protein synthesis and has an affinity for N-acetyl-D-galactosamine. RCA120, also referred to as RCAI or RCL (I + II), is an agglutinin and has an affinity for terminal β-D-galactosyl residues.
Conjugates are prepared from affinity purified agglutinin RCA120.
Lectin from Ricinus communis (castor bean) Agglutinin RCA120 is used to study cell surface glycoproteins the separation of glycoproteins with Galβ-terminating oligosaccharide chains from glycolipids.[2] Lectin agglutinates cells and precipitates polysaccharides and glycoproteins.[1]

Physical form

Solution in 0.005 M sodium phosphate and 0.2 M sodium chloride, pH 7.2, containing 0.1% sodium azide

Preparation Note

Purified by affinity chromatography using the method of Lin and Li.[5]

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 7.2, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes after 1 hour incubation at 25 °C.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Specificities of Ricinus communis agglutinin 120 interaction with sulfated galactose
Wang Y, et al.
Febs Letters, 585(24), 3927-3934 (2011)
T T Lin et al.
European journal of biochemistry, 105(3), 453-459 (1980-04-01)
Two toxin ricins and two agglutinins have been purified from the seeds of Ricinus communis by an improved procedure based on ion-exchange chromatography and gel filtration. One of the two purified ricins binds to Sepharose 4B while the other does
Identification of N-glycosites in chicken egg white proteins using an omics strategy
Geng F, et al.
Journal of Agricultural and Food Chemistry, 65(26), 5357-5364 (2017)
The Lectins: Properties, Functions, and Applications in Biology and Medicine (2012)
A photo-cleavable biotin affinity tag for the facile release of a photo-crosslinked carbohydrate-binding protein
Chang TC, et al.
Bioorganic & Medicinal Chemistry, 24(6), 1216-1224 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service