Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L6795

Sigma-Aldrich

LY311727

≥98% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27N2O5P
CAS Number:
Molecular Weight:
430.43
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥15 mg/mL

originator

Eli Lilly

storage temp.

2-8°C

SMILES string

CCc1c(CC(N)=O)c2cc(OCCCP(O)(O)=O)ccc2n1Cc3ccccc3

InChI

1S/C22H27N2O5P/c1-2-20-19(14-22(23)25)18-13-17(29-11-6-12-30(26,27)28)9-10-21(18)24(20)15-16-7-4-3-5-8-16/h3-5,7-10,13H,2,6,11-12,14-15H2,1H3,(H2,23,25)(H2,26,27,28)

InChI key

OPWQYOUZRHDKBR-UHFFFAOYSA-N

Biochem/physiol Actions

LY311727 is an orally active; potent secretory Phospholipase A2 (sPLA2; Group IIa) inhibitor.

Features and Benefits

This compound is featured on the Phospholipase A2 page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shulin Hou et al.
Scientific reports, 7(1), 10815-10815 (2017-09-09)
Secreted phospholipases A
Deepti Dabral et al.
Cells, 8(4) (2019-04-17)
The fundamental molecular mechanism underlying the membrane merger steps of regulated exocytosis is highly conserved across cell types. Although involvement of Phospholipase A₂ (PLA₂) in regulated exocytosis has long been suggested, its function or that of its metabolites-a lyso-phospholipid and

Articles

Phospholipase A2 (PLA2) enzymes hydrolyze glycerophospholipids, yielding arachidonic acid; inhibition targets eicosanoid biosynthesis.

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service