Skip to Content
MilliporeSigma
All Photos(1)

Documents

L6543

Sigma-Aldrich

Lacto-N-neo-tetraose

synthetic, ≥85% (HPLC)

Synonym(s):

β-D-Gal-(1→4)-β-D-GlcNAc-(1→3)-β-D-Gal-(1→4)-D-Glc, LNnT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H45NO21
CAS Number:
Molecular Weight:
707.63
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥85% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

CC(=O)NC1C(O)C(OC2OC(CO)C(O)C(O)C2O)C(CO)OC1OC3C(O)C(CO)OC(OC(C(O)CO)C(O)C(O)C=O)C3O

InChI

1S/C26H45NO21/c1-7(33)27-13-17(39)22(47-25-19(41)18(40)15(37)10(4-30)43-25)12(6-32)45-24(13)48-23-16(38)11(5-31)44-26(20(23)42)46-21(9(35)3-29)14(36)8(34)2-28/h2,8-26,29-32,34-42H,3-6H2,1H3,(H,27,33)

InChI key

RBMYDHMFFAVMMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Behrouz Farhadihosseinabadi et al.
Scientific reports, 10(1), 997-997 (2020-01-24)
Lacto-n-neotatraose (LNnT) oligosaccharide shows properties such as anti-inflammatory, type 2 immune response induction, induced angiogenesis, and anti-bacterial effects. Here, we hypothesized that the application of LnNT in the skin full-thickness wound can accelerate the healing process through its anti-inflammatory effect
Julie Elisabeth Heggelund et al.
International journal of molecular sciences, 20(3) (2019-02-10)
Diarrhea caused by enterotoxigenic Escherichia coli (ETEC) is one of the leading causes of mortality in children under five years of age and is a great burden on developing countries. The major virulence factor of the bacterium is the heat-labile
Stephen A Fleming et al.
Frontiers in neuroscience, 14, 770-770 (2020-09-10)
Human milk contains a unique profile of oligosaccharides (OS) and preliminary evidence suggests they impact brain development. The objective of this study was to assess the impact of bovine and/or human milk oligosaccharides (HMO) (2'-fucosyllactose and Lacto-N-neotetraose) on cognition, brain
Alexandra S Tauzin et al.
Microbiome, 8(1), 141-141 (2020-10-03)
Despite the importance of the mucosal interface between microbiota and the host in gut homeostasis, little is known about the mechanisms of bacterial gut colonization, involving foraging for glycans produced by epithelial cells. The slow pace of progress toward understanding
Liana Steeghs et al.
Cellular microbiology, 8(2), 316-325 (2006-01-31)
Neisseria meningitidis lipopolysaccharide (LPS) has been identified as a major determinant of dendritic cell (DC) function. Here we report that one of a series of meningococcal mutants with defined truncations in the lacto-N-neotetraose outer core of the LPS exhibited unique

Articles

O-Linked glycoproteins are usually large proteins with a molecular mass of >200 kDa. Glycosylation generally occurs in high-density clusters and may represent as much as 50-80% of the overall mass.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service