Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L6271

Sigma-Aldrich

L-Leucine allyl ester p-toluenesulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)COOCH2CH=CH2 · C7H8O3S
CAS Number:
Molecular Weight:
343.44
Beilstein:
5201666
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)S(O)(=O)=O.CC(C)C[C@H](N)C(=O)OCC=C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service