Skip to Content
MilliporeSigma
All Photos(1)

Documents

L5756

Sigma-Aldrich

Lithocholic acid 3-sulfate disodium salt hydrate

Synonym(s):

3α-Hydroxy-5β-cholan-24-oic acid 3-sulfate, Sulfolithocholic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38Na2O6S · xH2O
CAS Number:
Molecular Weight:
500.60 (anhydrous basis)
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

Assay

≥95% (TLC)

storage temp.

−20°C

SMILES string

[Na+].[Na+].[H]O[H].[H][C@]12CC[C@@]3([H])[C@]4([H])CC[C@H]([C@H](C)CCC([O-])=O)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CC[C@H](C2)OS([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Takikawa et al.
Digestive diseases and sciences, 40(8), 1792-1797 (1995-08-01)
Biliary excretion of lithocholate-3-sulfate is markedly impaired in EHBR. To examine the mechanism of biliary lithocholate-3-sulfate excretion in EHBR, the effects of colchicine treatment, a vesicular transport inhibitor, and infusion of taurocholate and organic anions were studied in EHBR and
Cholic acid binding by anionic glutathione-S-transferase from human liver cytosol.
N R Pattinson
Biochemical and biophysical research communications, 102(1), 403-410 (1981-09-16)
K Finni et al.
Acta paediatrica Scandinavica, 72(2), 215-218 (1983-03-01)
The concentrations of the two secondary bile acids (deoxycholic (DCA) and sulpholithocholic (SLCA) acid) were determined by radioimmunoassays in the serum of infants and children at ages ranging from 1 hour to 15 years. The same bile acids were measured
D G Oelberg et al.
The American journal of physiology, 247(1 Pt 1), G112-G115 (1984-07-01)
Lithocholic acid (LCA) and its sulfate (LCS) and glucuronide (LCG) derivatives are potent cholestatic agents. During the course of LCG-induced cholestasis in rats, calcium (Ca) salts of LCG precipitate in bile. To characterize the affinity of bile salts for Ca
N Tanida et al.
Cancer research, 49(5), 1178-1181 (1989-03-01)
Effects of p.o. administration of sulfolithocholic acid disodium salt (SLCNa) and lithocholic acid sodium salt (LCNa) on N-methyl-N-nitrosourea (MNU)-induced colonic tumorigenesis were studied in conventional rats. Female F344 rats received either 0.5 ml of distilled water (DW) alone or DW

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service