Skip to Content
MilliporeSigma
All Photos(1)

Documents

L4634

Sigma-Aldrich

Leu-Leu acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24N2O3 · C2H4O2
CAS Number:
Molecular Weight:
304.38
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

form

powder

color

white

storage temp.

−20°C

SMILES string

CC(O)=O.CC(C)C[C@H](N)C(=O)N[C@@H](CC(C)C)C(O)=O

InChI

1S/C12H24N2O3.C2H4O2/c1-7(2)5-9(13)11(15)14-10(12(16)17)6-8(3)4;1-2(3)4/h7-10H,5-6,13H2,1-4H3,(H,14,15)(H,16,17);1H3,(H,3,4)/t9-,10-;/m0./s1

InChI key

JEUHGRPWUPRNDP-IYPAPVHQSA-N

Amino Acid Sequence

Leu-Leu

Biochem/physiol Actions

Leucylleucine (Leu-Leu) may be used to study the functionality of dileucine motifs such as the motif responsible for internalization and targeting of vesicular acetylcholine transporter and clathrin adaptors AP-1 and AP-2. Leucylleucine may be used to study growth supplement requirements in vitro.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A van Boven et al.
Biochimie, 70(4), 535-542 (1988-04-01)
Different strains of Lactococcus lactis ssp. cremoris hydrolyze peptides at different rates while the cell-free extracts of these strains all show the same or much higher rates of hydrolysis. These observations indicate that the uptake of peptides is the rate-limiting
M Plauth et al.
Clinical science (London, England : 1979), 82(3), 283-290 (1992-03-01)
1. We assessed the efficacy of nitrogen absorption from luminal L-leucine (1.2 and 12 mmol/l) and from isonitrogenous L-leucyl-L-leucine (0.6 and 6.0 mmol/l) in a preparation of vascularly and luminally perfused rat small intestine by measuring luminal leucyl-leucine disappearance and
Lesley Colgan et al.
Traffic (Copenhagen, Denmark), 8(5), 512-522 (2007-04-25)
Efficient cholinergic transmission requires accurate targeting of vesicular acetylcholine transporter (VAChT) to synaptic vesicles (SVs). However, the signals that regulate this vesicular targeting are not well characterized. Although previous studies suggest that the C-terminus of the transporter is required for
O Valdenaire et al.
Journal of cell science, 112 Pt 18, 3115-3125 (1999-08-27)
Endothelin-converting enzyme (ECE-1) is a type II integral membrane protein which plays a key role in the biosynthetic pathway of the vasoconstricting endothelins. Three ECE-1 isoforms, differing by their N-terminal cytoplasmic tails, are generated from a single gene. When expressed
Pedro A Aredes Fernández et al.
Current microbiology, 49(5), 361-365 (2004-10-16)
Oenococcus oeni has numerous amino acid requirements for growth and dipeptides could be important for its nutrition. In this paper the individual or combined effect of dipeptides on growth of O. oeni X2L in synthetic media deficient in one or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service