Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L4376

Sigma-Aldrich

α-Lobeline hydrochloride

~95% (HPLC), powder

Synonym(s):

2-[6-(2-Hydroxy-2-phenylethyl)-1-methyl-2-piperidinyl]-1-phenylethanone hydrochloride, (−)-Lobeline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27NO2 · HCl
CAS Number:
Molecular Weight:
373.92
Beilstein:
3920196
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

~95% (HPLC)

form

powder

color

white to off-white

solubility

H2O: 25 mg/mL

SMILES string

Cl.CN1[C@@H](CCC[C@@H]1CC(=O)c2ccccc2)C[C@H](O)c3ccccc3

Gene Information

human ... SLC18A2(6571)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Agonist at high-affinity neuronal nicotinic acetylcholine receptors; respiratory stimulant. Inhibits synaptosomal accumulation of dopamine by interaction with the tetrabenazine binding site of the vesicular monoamine transporter (VMAT2). Blocks amphetamine-induced dopamine release.

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service