Skip to Content
MilliporeSigma
All Photos(1)

Documents

L4015

Sigma-Aldrich

Lipoteichoic acid from Enterococcus hirae

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

biological source

bacterial (Enterococcus hirae)

Quality Level

form

lyophilized powder

shipped in

ambient

storage temp.

2-8°C

General description

the bacterial source Streptococcus faecalis has been renamed Enterococcus hirae

Biochem/physiol Actions

Lipoteichoic acid (LTA) is a complex component of cell walls of Gram-positive bacteria that are involved in a wide range of cell processes such as the stimulation of immune responses and cell signaling pathways. LTA differs between species of gram-positive bacteria. Lipoteichoic acid from Streptococcus faecalis may be used to compare its structure, immunogenicity and functions versus other bacterial LTAs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zdeněk Zídek et al.
Nitric oxide : biology and chemistry, 23(4), 300-310 (2010-09-21)
Lipoteichoic acid (LTA) is a structural component of the cell walls of Gram-positive bacteria. Similar to lipopolysaccharide (LPS) which is expressed in Gram-negative bacteria, LTA exhibits immunostimulatory properties. Frequently observed positive response of LTA in the Limulus amebocyte lysate (LAL)
G L Card et al.
Journal of leukocyte biology, 56(6), 723-728 (1994-12-01)
The relative activities of lipoteichoic acid (LTA) from four Gram-positive bacteria were compared to different lipopolysaccharide (LPS) preparations for activation of arachidonic acid metabolism in mouse peritoneal macrophages. Total eicosanoid was determined in cultures labeled with [3H]-arachidonic acid. Prostaglandin E2
Tetsuaki Ishikawa et al.
Cell host & microbe, 13(4), 477-488 (2013-04-23)
Various C-type lectin receptors (CLRs), including Mincle and Dectin-2, function as pattern recognition receptors and play a central role in immunity to fungal pathogens. However, the precise structures of the CLR ligands in various pathogenic fungi have yet to be
Anton Steen et al.
The Journal of biological chemistry, 278(26), 23874-23881 (2003-04-10)
The C-terminal region (cA) of the major autolysin AcmA of Lactococcus lactis contains three highly similar repeated regions of 45 amino acid residues (LysM domains), which are separated by nonhomologous sequences. The cA domain could be deleted without destroying the
Christoph Thiemermann
Microbes and infection, 4(9), 927-935 (2002-07-11)
The cell wall of Gram-positive bacteria contains lipoteichoic acid (LTA) and peptidoglycan (PepG), which synergise to cause shock and organ failure in animals, and to activate human blood to release proinflammatory cytokines. The structural elements within LTA and PepG that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service