Skip to Content
MilliporeSigma
All Photos(1)

Documents

L3502

Sigma-Aldrich

D-Leu-Gly

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2O3
CAS Number:
Molecular Weight:
188.22
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CC(C)C[C@@H](N)C(=O)NCC(O)=O

InChI

1S/C8H16N2O3/c1-5(2)3-6(9)8(13)10-4-7(11)12/h5-6H,3-4,9H2,1-2H3,(H,10,13)(H,11,12)/t6-/m1/s1

InChI key

LESXFEZIFXFIQR-ZCFIWIBFSA-N

Biochem/physiol Actions

D-Leucylglycine (D-Leu-Gly) is used to study the kinetics and speciation of copper(II)- cis, cis-1,3,5-triaminocyclohexane complex-promoted hydrolysis of dipeptides and the specificity of cleavage of leucine dipeptides by peptidases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Fujii et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 7(7-8), 843-851 (2002-08-31)
The hydrolysis of glycylglycine (GylGly), glycyl-L-leucine (GlyLeu), L-leucylglycine (LeuGly) and glycyl-DL-serine (GlySer) promoted by a copper(II)- cis, cis-1,3,5-triaminocyclohexane complex [Cu(II)TACH] was investigated at 70 degrees C and pH 7-10, using HPLC. The observed pseudo-first-order rate constants (k(obs)) and rate enhancing
N Khelifa et al.
Bioorganic & medicinal chemistry letters, 9(1), 109-112 (1999-02-17)
The ability of Clostridium butyricum cultures to hydrolyze three L-leucine-containing dipeptides (Leu-Leu, Leu-Gly and Gly-Leu) in a synthetic minimal medium is demonstrated by using gas chromatography coupled with mass spectrometry. The 13C nuclear magnetic resonance and a labeled dipeptide L-[1-13C]Leu-Gly
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service