Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L3150

Sigma-Aldrich

Monoclonal Anti-LEF-1 (Transactivation Domain 236-242) antibody produced in mouse

~1 mg/mL, clone 3A12, purified immunoglobulin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A12, monoclonal

mol wt

antigen 55-57 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1-5 μg/mL

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... LEF1(51176)

Immunogen

synthetic peptide corresponding to amino acids 236-242 (DTSMSRF) of human LEF-1.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.5, containing 0.08% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service