Skip to Content
MilliporeSigma
All Photos(1)

Documents

L2792

Sigma-Aldrich

LCK, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

YT16, p56lck, pp58lck

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

180-244 nmol/min·mg

mol wt

~84 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... LCK(3932)

Biochem/physiol Actions

LCK (proto-oncogene) belongs to the Src family of kinase and is mainly expressed in T cells. It is important for T cell development and homeostasis. LCK is upregulated in acute and chronic leukemias, colorectal and prostate cancers.
Tyrosine kinase that activates T-lymphocytes.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Fischer et al.
Biochemical and biophysical research communications, 143(3), 819-826 (1987-03-30)
High levels of tyrosine kinase activity have been detected in the murine lymphoma LSTRA (p56). The functional domains of this kinase have been studied by the use of antibodies generated against peptides from the amino terminal region and from the
A Veillette et al.
Oncogene research, 1(4), 357-374 (1987-09-01)
The tyrosine kinase p56lck is felt to be expressed solely in cells and tissues of lymphoid lineage. We have observed in the particulate fractions of colon carcinoma cells the presence of a 56 kilodalton (kD) alkali resistant phosphoprotein containing phosphotyrosine.
Shahrooz Vahedi et al.
Oncology reports, 34(1), 43-50 (2015-05-23)
Overexpression and hyperactivation of lymphocyte-specific protein tyrosine kinase (Lck) have been associated with leukemia development. We previously showed that, other than its known function as a cytoplasmic signal transducer, Lck also acts as a nuclear transcription factor in mouse leukemic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service