Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L2650

Sigma-Aldrich

Lectin from Glycine max (soybean)

peroxidase conjugate, lyophilized powder

Synonym(s):

SBA, Soybean agglutinin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

Glycine max (soybean)

conjugate

peroxidase conjugate

Assay

85.00-100.00% (Warburg-Christian)

form

lyophilized powder

potency

<80 μg per mL agglutination activity

peroxidase activity

25-100 units/mg protein

composition

Protein, ~95% modified Warburg-Christian

technique(s)

western blot: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

soybean ... LE1(100818710)

Looking for similar products? Visit Product Comparison Guide

Application

General Western Blot Protocol:
  • Glycoprotein sample size: 500ng
  • Lectin Concentration: 0.1ug/ml

  1. Load samples at 500 ng of glycoprotein per lane
  2. Run 4-20% Bis-Tris SDS page gel
  3. Transfer gel to a PVDF membrane
  4. Block membrane for 1 hr at RT with RIPA buffer (R0278 Sigma)
  5. Incubate HRP lectin at 0.1ug/ml with RIPA buffer for 2 hours at RT
  6. Wash membrane 5 x 5 minutes with 25ml RIPA buffer
  7. Detect using chemiluminescent substrate (CPS1-120)

Biochem/physiol Actions

SBA is not blood group specific but has an affinity for N-acetyl-D-galactosamine.

Unit Definition

One unit will form 1 mg purpurogallin in 20 sec from pyrogallol at pH 6.0, at 20 °C.

Physical form

Contains sodium citrate

Preparation Note

Conjugates are prepared from affinity purified lectin.
Prepared from peroxidase type VI (P8375) using a modification of the method of O′Sullivan, which favors low molecular weight conjugates. Repurified after conjugation by affinity chromatography.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg per mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group A erythrocytes after 1 hr incubation at 25 °C.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Biel et al.
Archives of toxicology, 96(6), 1815-1827 (2022-04-17)
Human Precision-cut intestinal slices (hPCIS) are used to study intestinal physiology, pathophysiology, drug efficacy, toxicology, kinetics, and metabolism. However, the use of this ex vivo model is restricted to approximately a 24 h timeframe because of declining viability of the hPCIS
Carin Biel et al.
Toxicology in vitro : an international journal published in association with BIBRA, 69, 104974-104974 (2020-08-24)
Precision-cut intestinal slices (PCIS) are used to study intestinal (patho)physiology, drug efficacy, toxicity, transport and metabolism ex vivo. One of the factors that limit the use of PCIS is a relatively short life-span. Moreover, culture-induced changes in cellular composition of
Jonathan Welch et al.
Regulatory toxicology and pharmacology : RTP, 125, 105022-105022 (2021-08-02)
The aim of the study was to use multiple in vitro assays to assess the effects of a model irritant, sodium dodecyl sulphate (SDS) (≤10 mM (0.29 %, w/v)), on an in vitro model of the airway, MucilAir™. The use of MucilAir™
Tomás Edreira et al.
eLife, 9 (2020-10-27)
In fission yeast, the septation initiation network (SIN) ensures temporal coordination between actomyosin ring (CAR) constriction with membrane ingression and septum synthesis. However, questions remain about CAR regulation under stress conditions. We show that Rgf1p (Rho1p GEF), participates in a
Daniel Sanchez-Guzman et al.
Scientific reports, 11(1), 6621-6621 (2021-03-25)
The human bronchial epithelium is the first line of defense against atmospheric particles, pollutants, and respiratory pathogens such as the novel SARS-CoV-2. The epithelial cells form a tight barrier and secrete proteins that are major components of the mucosal immune

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service