Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L1414

Sigma-Aldrich

Labiase from Streptomyces fulvissimus TU-6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

form

powder

specific activity

≥10.0 units/g solid

storage temp.

2-8°C

Application

Labiase from Streptomyces fulvissimus is an enzyme preparation useful for the lysis of many Gram-positive bacteria such as Lactobacillus, Aerococcus and, Streptococcus.
Labiase contains β-N-acetyl-D-glucosaminidase and lysozyme activity.
pH Optimum for activity: pH ~4
pH Optimum for stability: pH 4-8

Unit Definition

One unit will release 1 micromole of p-nitrophenol from p-nitrophenyl N-acetyl-beta-D-glucosaminide per minute at pH 4.0 at 37 deg C.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Niwa et al.
Journal of microbiological methods, 61(2), 251-260 (2005-02-22)
The lytic activity of labiase and achromopeptidase for bacterial DNA/RNA extraction were compared. Rapid lysis of many bacterial strains was observed with labiase followed by SDS treatment. Both labiase and achromopeptidase showed high lytic activity against bacterial strains with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service