Skip to Content
MilliporeSigma
All Photos(4)

Documents

L1000

Sigma-Aldrich

Lithium D-lactate

95-100%

Synonym(s):

(R)-2-Hydroxypropionic acid lithium salt, D-Lactic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CO2Li
CAS Number:
Molecular Weight:
96.01
Beilstein:
4823618
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

95-100%

storage temp.

−20°C

SMILES string

[Li+].C[C@@H](O)C([O-])=O

InChI

1S/C3H6O3.Li/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m1./s1

InChI key

GKQWYZBANWAFMQ-HSHFZTNMSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Lactic acid is used as a reagent in organic synthesis (in the manufacture of adhesives). It is used in the leather, textile, and tanning industries. It may be used as a plasticizer, a catalyst, or an acidifying agent. Lactic acid has even been used as a flavoring agent in the manufacture of tobacco products.

Biochem/physiol Actions

In animals, lactic acid is a metabolic compound produced by proliferating cells and during anaerobic conditions such as strenuous exercise. Lactic acid can be oxidized back to pyruvate or converted to glucose via gluconeogenesis. Lactic acid is preferentially metabolized by neurons in several mammal species and during early brain development. D-lactate was utilized four times more slowly than L-lactate, but both isomers are absorbed at the same rate from the intestine.

Analysis Note

Assayed enzymatically using D(−)lactic dehydrogenase

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heverton Leandro Carneiro Dutra et al.
PLoS pathogens, 16(10), e1008996-e1008996 (2020-10-14)
Wolbachia is an intracellular bacterial symbiont of arthropods notorious for inducing many reproductive manipulations that foster its dissemination. Wolbachia affects many aspects of host biology, including metabolism, longevity and physiology, being described as a nutrient provisioning or metabolic parasite, depending
Chien-Ming Chen et al.
Journal of pharmaceutical and biomedical analysis, 116, 150-155 (2015-08-13)
In this study, we aimed to develop a new enzymatic assay system of d-lactate with good precision, accuracy, and sensitivity for the determination of D-lactate concentrations in rat serum. D-Lactate dehydrogenase (D-LDH) was utilized to catalyze D-lactate and NAD(+) to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service